Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025082.3 | NP_079358.3 | 8 | 1020 | ||
XM_006721277.2 | XP_006721340.1 | 6 | 875 | |||
XM_011523344.1 | XP_011521646.1 | 7 | 997 | |||
XM_017023712.1 | XP_016879201.1 | 6 | 874 | |||
XM_017023713.1 | XP_016879202.1 | 7 | 1001 | |||
XM_017023714.1 | XP_016879203.1 | 7 | 962 | |||
XM_017023715.1 | XP_016879204.1 | 4 | 302 | |||
XM_017023716.1 | XP_016879205.1 | 7 | 1001 | |||
GenBank mRNA | AK023173.1 | - | 5 | 509 | ||
AK055237.1 | - | 4 | 677 | |||
AK056097.1 | - | 8 | 1020 | |||
AK125366.1 | - | 3 | 751 | |||
AK291527.1 | - | 5 | 540 | |||
AK297570.1 | - | 4 | 409 | |||
AK301621.1 | - | 5 | 423 | |||
BC007642.1 | - | 5 | 463 | |||
BC015202.2 | - | 5 | 471 | 3878214 | ||
BX648617.1 | - | 2 | 149 |