Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033580.2 | NP_001028752.1 | 7 | 1418 | ||
NM_001163735.1 | NP_001157207.1 | 7 | 1418 | |||
NM_025109.5 | NP_079385.2 | 7 | 1418 | |||
XM_011525286.1 | XP_011523588.1 | 8 | 967 | |||
XM_011525287.1 | XP_011523589.1 | 8 | 967 | |||
XM_011525290.2 | XP_011523592.1 | 8 | 967 | |||
XM_017025157.1 | XP_016880646.1 | 8 | 941 | |||
XM_017025158.1 | XP_016880647.1 | 7 | 874 | |||
XM_017025159.1 | XP_016880648.1 | 8 | 967 | |||
XM_017025160.1 | XP_016880649.1 | 8 | 967 | |||
XM_017025161.1 | XP_016880650.1 | 7 | 829 | |||
GenBank mRNA | AB209035.1 | - | 7 | 1376 | ||
AK026518.1 | - | 7 | 829 | |||
AK293903.1 | - | 7 | 1418 | |||
AK299802.1 | - | 4 | 455 | |||
AK304073.1 | - | 7 | 769 | |||
BC008900.2 | - | 7 | 809 | 3629870 |