Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALPK1 SIRNA Assays ›
Gene Symbol:
ALPK1
Gene Name:
alpha kinase 1
Gene Aliases:
8430410J10Rik, LAK
Chromosome Location:
Chr.4: 112286420 - 112442618 on Build GRCh38
UniGene ID:
Hs.652825
Species:
Human
siRNA ID
HSS188397
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALPK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102406.1
NP_001095876.1
11
2960
NM_001253884.1
NP_001240813.1
10
2805
NM_025144.3
NP_079420.3
11
3012
XM_005263245.4
XP_005263302.1
11
3012
XM_005263246.4
XP_005263303.1
11
2935
XM_006714326.3
XP_006714389.1
10
2952
XM_017008633.1
XP_016864122.1
11
3030
XM_017008634.1
XP_016864123.1
10
2998
XM_017008635.1
XP_016864124.1
11
2953
XM_017008636.1
XP_016864125.1
12
3106
XM_017008637.1
XP_016864126.1
11
2981
XM_017008638.1
XP_016864127.1
11
3119
XM_017008639.1
XP_016864128.1
12
3054
XM_017008640.1
XP_016864129.1
11
2929
XM_017008641.1
XP_016864130.1
11
3067
XM_017008642.1
XP_016864131.1
11
3030
XM_017008643.1
XP_016864132.1
10
2970
XM_017008644.1
XP_016864133.1
8
6163
XM_017008645.1
XP_016864134.1
10
2952
XM_017008646.1
XP_016864135.1
10
2907
XM_017008647.1
XP_016864136.1
10
2881
XM_017008648.1
XP_016864137.1
10
2829
XM_017008649.1
XP_016864138.1
6
2205
XM_017008650.1
XP_016864139.1
11
4615
XM_017008651.1
XP_016864140.1
11
2925
GenBank mRNA
AB040960.1
-
1
1405
AK131090.1
-
7
3221
AK304708.1
-
10
2768
AY044164.1
-
9
2815
BX648798.1
-
11
2903
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
protein phosphorylation
mitophagy in response to mitochondrial depolarization
xenophagy
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline