Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025165.2 | NP_079441.1 | 1, 2 | 545 | ||
GenBank mRNA | AF276512.1 | - | 1, 2 | 142 | ||
AK026290.1 | - | 1, 2 | 227 | |||
AK126384.1 | - | 1, 2 | 784 | |||
BC006548.2 | - | 1, 2 | 196 | 2821972 | ||
BC019293.1 | - | 1, 2 | 196 | 2821972 | ||
BC062349.1 | - | 1 | 34 | |||
CR457358.1 | - | 1, 2 | 142 |