Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025184.3 | NP_079460.2 | 13 | 2098 | ||
XM_006724562.2 | XP_006724625.1 | 12 | 1811 | |||
GenBank mRNA | AK026254.1 | - | 3 | 372 | ||
AK026496.1 | - | 13 | 1952 | |||
AK291187.1 | - | 13 | 2239 | |||
AK292967.1 | - | 13 | 2082 | |||
BC031039.1 | - | 13 | 2093 | 5272398 | ||
CR749363.1 | - | 8 | 1761 |