Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320540.1 | NP_001307469.1 | 5 | 375 | ||
NM_001320541.1 | NP_001307470.1 | 5 | 416 | |||
NM_001320542.1 | NP_001307471.1 | 5 | 401 | |||
NM_001320543.1 | NP_001307472.1 | 4 | 332 | |||
NM_025187.4 | NP_079463.2 | 4 | 431 | |||
XM_006721284.3 | XP_006721347.1 | 5 | 399 | |||
XM_011523350.2 | XP_011521652.1 | 4 | 539 | |||
XM_017023729.1 | XP_016879218.1 | 5 | 410 | |||
XM_017023730.1 | XP_016879219.1 | 5 | 413 | |||
XM_017023731.1 | XP_016879220.1 | 5 | 411 | |||
GenBank mRNA | AF176088.1 | - | 5 | 331 | ||
AK022138.1 | - | 3 | 200 | |||
AL540304.3 | - | 5 | 310 | |||
BC004556.2 | - | 4 | 411 | 3957147 | ||
CB997191.1 | - | 4 | 290 | |||
HY121038.1 | - | 4 | 432 | |||
HY157930.1 | - | 5 | 401 |