Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005436.4 | NP_005427.2 | 8 | 1443 | ||
GenBank mRNA | AA112388.1 | - | 1 | 88 | ||
AK056848.1 | - | 2 | 1263 | |||
AK292593.1 | - | 8 | 1443 | |||
BC036757.2 | - | 8 | 1282 | 5549623 | ||
BC064391.1 | - | 8 | 1269 | 5745698 | ||
S72869.1 | - | 8 | 1247 |