Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143757.1 | NP_001137229.1 | 2 | 464 | ||
NM_001143758.1 | NP_001137230.1 | 2 | 311 | |||
NM_001143759.1 | NP_001137231.1 | 2 | 311 | |||
NM_030626.2 | NP_085129.1 | 2 | 311 | |||
XM_006717983.2 | XP_006718046.1 | 2 | 624 | |||
XM_006717984.2 | XP_006718047.1 | 2 | 311 | |||
XM_011540208.1 | XP_011538510.1 | 2 | 394 | |||
XM_011540209.1 | XP_011538511.1 | 2 | 311 | |||
XM_017016690.1 | XP_016872179.1 | 2 | 311 | |||
XM_017016691.1 | XP_016872180.1 | 2 | 311 | |||
XM_017016692.1 | XP_016872181.1 | 2 | 311 | |||
XM_017016693.1 | XP_016872182.1 | 2 | 311 | |||
XM_017016694.1 | XP_016872183.1 | 2 | 257 | |||
GenBank mRNA | AB051461.1 | - | 2 | 221 | ||
AK097418.1 | - | 2 | 422 | |||
AK295953.1 | - | 2 | 228 | |||
BC050097.1 | - | 2 | 266 | 5273395 | ||
DA203895.1 | - | 2 | 464 | |||
DA658738.1 | - | 2 | 311 |