Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032548.3 | NP_115937.1 | 9 | 906 | ||
NM_172027.2 | NP_742024.1 | 9 | 895 | |||
NR_033429.1 | 9 | 860 | ||||
XM_006713769.3 | XP_006713832.1 | 9 | 929 | |||
XM_011513213.1 | XP_011511515.1 | 9 | 829 | |||
XM_017007285.1 | XP_016862774.1 | 8 | 758 | |||
XM_017007286.1 | XP_016862775.1 | 8 | 740 | |||
XM_017007287.1 | XP_016862776.1 | 9 | 866 | |||
XM_017007288.1 | XP_016862777.1 | 8 | 799 | |||
GenBank mRNA | AB053324.1 | - | 9 | 885 | ||
AB053325.1 | - | 9 | 874 | |||
AB053326.1 | - | 9 | 839 | |||
AF297986.1 | - | 9 | 852 | |||
AF447886.1 | - | 8 | 999 | |||
AK090422.1 | - | 3 | 1742 | |||
AK315148.1 | - | 9 | 875 | |||
BC011858.2 | - | 9 | 893 | 4309664 |