Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025218.3 | NP_079494.1 | 3 | 544 | ||
NR_133659.1 | 3 | 1892 | ||||
XM_017011322.1 | XP_016866811.1 | 3 | 544 | |||
GenBank mRNA | AB052907.1 | - | 3 | 468 | ||
AF304377.1 | - | 3 | 468 | |||
AF346416.1 | - | 3 | 468 | |||
AF346595.1 | - | 3 | 438 | |||
AF425265.1 | - | 3 | 455 | |||
AK292519.1 | - | 3 | 544 | |||
AK316582.1 | - | 3 | 533 | |||
BC035416.1 | - | 3 | 511 | 4822311 | ||
KT591165.1 | - | 3 | 1859 |