Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124756.2 | NP_001118228.1 | 7, 8 | 1047 | ||
GenBank mRNA | BX451955.2 | - | 2, 3 | 208 |