Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003476.4 | NP_003467.1 | 5 | 608 | ||
GenBank mRNA | AF121260.1 | - | 4 | 498 | ||
AK313289.1 | - | 4 | 433 | |||
BC005900.2 | - | 3 | 398 | 4247505 | ||
BC024010.1 | - | 5 | 611 | 4826902 | ||
BC057221.1 | - | 4 | 431 | 4337632 | ||
U20324.1 | - | 4 | 414 | |||
U49837.1 | - | 4 | 426 | |||
U72899.1 | - | 4 | 414 |