Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001851.2 | NP_001001851.1 | 9 | 1395 | ||
NM_030569.6 | NP_085046.5 | 9 | 1395 | |||
NM_032817.5 | NP_116206.4 | 5 | 730 | |||
XM_011519713.2 | XP_011518015.1 | 10 | 1470 | |||
XM_011519714.2 | XP_011518016.1 | 10 | 1470 | |||
GenBank mRNA | AB075833.1 | - | 6 | 936 | ||
AK027375.1 | - | 9 | 1332 | |||
AK027831.1 | - | 6 | 914 | |||
AK027849.1 | - | 4 | 579 | |||
AK075381.1 | - | 9 | 1368 | |||
AL833203.1 | - | 9 | 1240 | |||
AY238437.1 | - | 9 | 1291 | |||
AY358426.1 | - | 9 | 1360 | |||
BC136439.1 | - | 9 | 1368 | |||
CR627109.1 | - | 4 | 5829 | |||
DA833541.1 | - | 4 | 579 |