Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001851.2 | NP_001001851.1 | 10 | 1708 | ||
NM_030569.6 | NP_085046.5 | 10 | 1708 | |||
NM_032817.5 | NP_116206.4 | 6 | 1043 | |||
XM_011519713.2 | XP_011518015.1 | 11 | 1783 | |||
XM_011519714.2 | XP_011518016.1 | 11 | 1783 | |||
GenBank mRNA | AB075833.1 | - | 7 | 1249 | ||
AF318347.1 | - | 2 | 593 | |||
AK027375.1 | - | 10 | 1645 | |||
AK027831.1 | - | 7 | 1227 | |||
AK027849.1 | - | 5 | 892 | |||
AK075381.1 | - | 10 | 1681 | |||
AL833203.1 | - | 10 | 1553 | |||
AY238437.1 | - | 10 | 1604 | |||
AY358426.1 | - | 10 | 1673 | |||
BC136439.1 | - | 10 | 1681 | |||
CR627109.1 | - | 5 | 6142 |