Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181900.2 | NP_871629.1 | 6 | 594 | ||
NR_135013.1 | 5 | 544 | ||||
XM_017022639.1 | XP_016878128.1 | 5 | 1109 | |||
GenBank mRNA | AF480304.1 | - | 6 | 510 | ||
AK026352.1 | - | 5 | 502 | |||
AK097024.1 | - | 4 | 1962 | |||
AK225479.1 | - | 5 | 502 | |||
AL137657.1 | - | 5 | 544 | |||
BC004365.1 | - | 6 | 536 | 3641640 |