Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029885.1 | NP_001025056.1 | 2, 3 | 532 | ||
XM_005244801.3 | XP_005244858.1 | 2, 3 | 645 | |||
XM_011542200.2 | XP_011540502.1 | 2, 3 | 541 | |||
GenBank mRNA | JN542538.1 | - | 1, 2 | 117 |