Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 39 | 4230 | ||
NM_130444.2 | NP_569711.2 | 39 | 4935 | |||
NM_130445.3 | NP_569712.2 | 40 | 3763 | |||
GenBank mRNA | AF018081.1 | - | 39 | 4249 | ||
AF018082.1 | - | 40 | 3728 | |||
AF184060.1 | - | 2 | 213 | |||
AF416592.1 | - | 2 | 222 | |||
AK098216.1 | - | 6 | 1853 | |||
AK130835.1 | - | 7 | 880 | |||
BC033715.1 | - | 28 | 2111 | |||
BC063833.1 | - | 40 | 3704 | 6181818 | ||
L22548.1 | - | 21 | 1713 |