Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005086.4 | NP_005077.2 | 1, 2 | 386 | ||
GenBank mRNA | AF016028.2 | - | 1, 2 | 286 | ||
AK304667.1 | - | 2, 3 | 280 | |||
AK314353.1 | - | 1, 2 | 375 | |||
AL136756.1 | - | 1, 2 | 370 | |||
BC062299.1 | - | 1, 2 | 366 | 6376868 | ||
EU433933.1 | - | 1, 2 | 286 | |||
X89105.1 | - | 1, 2 | 208 |