Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080423.3 | NP_001073892.3 | 3, 4 | 296 | ||
XM_011534141.2 | XP_011532443.1 | 3, 4 | 378 | |||
XM_011534142.2 | XP_011532444.1 | 3, 4 | 290 | |||
XM_011534143.2 | XP_011532445.1 | 3, 4 | 378 | |||
XM_011534146.2 | XP_011532448.1 | 3, 4 | 371 | |||
GenBank mRNA | AB051506.2 | - | 3, 4 | 282 | ||
AK074145.1 | - | 2, 3 | 273 | |||
AK095762.1 | - | 3, 4 | 296 |