Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030647.1 | NP_085150.1 | 16 | 2092 | ||
XM_011516587.2 | XP_011514889.1 | 10 | 1117 | |||
GenBank mRNA | AB051505.1 | - | 4 | 397 | ||
AK131497.1 | - | 16 | 2093 | |||
BX641017.1 | - | 8 | 872 | |||
BX649115.1 | - | 11 | 1359 |