Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305008.1 | NP_001291937.1 | 13 | 1484 | ||
NM_001305009.1 | NP_001291938.1 | 14 | 1572 | |||
NM_001305010.1 | NP_001291939.1 | 12 | 1292 | |||
NM_001305011.1 | NP_001291940.1 | 11 | 1249 | |||
NM_030650.2 | NP_085153.1 | 13 | 1479 | |||
NR_130941.1 | 15 | 1843 | ||||
XM_005246878.3 | XP_005246935.1 | 14 | 1498 | |||
XM_005246879.4 | XP_005246936.1 | 13 | 1426 | |||
XM_005246880.3 | XP_005246937.1 | 9 | 1116 | |||
XM_006712783.2 | XP_006712846.1 | 13 | 1621 | |||
XM_011511945.2 | XP_011510247.1 | 14 | 1830 | |||
XM_017005055.1 | XP_016860544.1 | 9 | 1024 | |||
GenBank mRNA | AB051502.1 | - | 13 | 1385 | ||
AK056532.1 | - | 13 | 1402 | |||
AK301947.1 | - | 15 | 1790 | |||
AK302833.1 | - | 12 | 1226 | |||
AL832947.1 | - | 10 | 1142 | |||
BC031530.1 | - | 2 | 332 | |||
BC105132.1 | - | 13 | 1409 | 8144135 | ||
BC105134.1 | - | 13 | 1409 | 8144137 | ||
BC110329.1 | - | 11 | 1178 | 5732809 | ||
BC143681.1 | - | 14 | 1502 | 9052195 | ||
BC143683.1 | - | 12 | 1222 | |||
CR936742.1 | - | 13 | 1428 |