Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280543.1 | NP_001267472.1 | 10 | 1160 | ||
NM_001280544.1 | NP_001267473.1 | 9 | 1459 | |||
NM_012074.4 | NP_036206.3 | 9 | 1016 | |||
XM_017021671.1 | XP_016877160.1 | 9 | 1766 | |||
XM_017021673.1 | XP_016877162.1 | 9 | 1356 | |||
XM_017021674.1 | XP_016877163.1 | 9 | 1545 | |||
GenBank mRNA | AK124946.1 | - | 1 | 51 | ||
AK126074.1 | - | 6 | 700 | |||
AK126933.1 | - | 9 | 1459 | |||
AK294425.1 | - | 10 | 1145 | |||
BC026305.1 | - | 9 | 1001 | |||
BC060801.1 | - | 9 | 1001 | 5260159 |