Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098725.1 | NP_001092195.1 | 3 | 445 | ||
NM_021966.2 | NP_068801.1 | 3 | 445 | |||
NR_049726.1 | 4 | 669 | ||||
XM_017021676.1 | XP_016877165.1 | 3 | 401 | |||
XM_017021677.1 | XP_016877166.1 | 3 | 401 | |||
GenBank mRNA | BC003574.1 | - | 3 | 357 | 3544215 | |
BC005831.2 | - | 3 | 357 | 2966563 | ||
BC014024.1 | - | 3 | 361 | 4308655 | ||
BG718395.1 | - | 3 | 450 | |||
BU602054.1 | - | 4 | 600 | |||
X82240.1 | - | 3 | 360 |