Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199671.1 | NP_001186600.1 | 3, 4 | 1190 | ||
NM_001199673.1 | NP_001186602.1 | 2, 3 | 375 | |||
NM_001219.4 | NP_001210.1 | 2, 3 | 375 | |||
GenBank mRNA | AF013759.1 | - | 2, 3 | 281 | ||
AK056338.1 | - | 3, 4 | 1078 | |||
AK074862.1 | - | 2, 3 | 325 | |||
AK312446.1 | - | 2, 3 | 318 | |||
BC013383.2 | - | 2, 3 | 274 | 3507961 | ||
BT006826.1 | - | 1, 2 | 223 | |||
BT020143.1 | - | 1, 2 | 223 | |||
CR457039.1 | - | 1, 2 | 223 | |||
CR541835.1 | - | 1, 2 | 223 | |||
DA023987.1 | - | 2, 3 | 375 | |||
DA703012.1 | - | 2, 3 | 288 | |||
HM002604.1 | - | 3, 4 | 1191 | |||
HM002606.1 | - | 2, 3 | 375 | |||
HM002607.1 | - | 2, 3 | 375 | |||
HM002610.1 | - | 2, 3 | 375 | |||
HM002611.1 | - | 2, 3 | 375 | |||
HM002615.1 | - | 3, 4 | 1191 | |||
U67280.1 | - | 2, 3 | 285 |