Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030790.4 | NP_110417.2 | 17 | 1910 | ||
GenBank mRNA | AF212247.1 | - | 17 | 1818 | ||
AF503339.1 | - | 17 | 1700 | |||
AK027596.1 | - | 17 | 1701 | |||
AK295228.1 | - | 9 | 846 | |||
AK299641.1 | - | 12 | 1144 | |||
AL833705.1 | - | 11 | 1016 | |||
BC006321.2 | - | 17 | 1683 | 4137934 | ||
BC024162.1 | - | 17 | 1744 | 4046297 |