Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL21A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318751.1 | NP_001305680.1 | 28 | 2682 | ||
NM_001318752.1 | NP_001305681.1 | 26 | 2563 | |||
NM_001318753.1 | NP_001305682.1 | 11 | 729 | |||
NM_001318754.1 | NP_001305683.1 | 11 | 727 | |||
NM_030820.3 | NP_110447.2 | 27 | 2608 | |||
NR_134849.1 | 27 | 2600 | ||||
NR_134850.1 | 28 | 2625 | ||||
NR_134851.1 | 27 | 2597 | ||||
XM_006715223.1 | XP_006715286.1 | 26 | 2738 | |||
XM_011514924.2 | XP_011513226.1 | 27 | 2551 | |||
XM_011514925.2 | XP_011513227.1 | 29 | 2721 | |||
XM_011514926.1 | XP_011513228.1 | 27 | 2479 | |||
XM_011514927.1 | XP_011513229.1 | 27 | 2433 | |||
GenBank mRNA | AF330693.1 | - | 26 | 2529 | ||
AF370383.1 | - | 15 | 864 | |||
AF414088.1 | - | 27 | 2578 | |||
AK096444.1 | - | 11 | 730 | |||
AK126586.1 | - | 28 | 2493 | |||
AK313398.1 | - | 27 | 2584 | |||
AL136624.1 | - | 27 | 2578 | |||
BC045597.1 | - | 27 | 2555 | 4814199 | ||
BC126108.1 | - | 27 | 2463 | 8991824 | ||
BC143862.1 | - | 27 | 2455 | |||
BC143863.1 | - | 28 | 2480 | |||
BC143864.1 | - | 27 | 2452 | |||
BC143865.1 | - | 28 | 2537 | 9052381 |