Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128309.1 | NP_001121781.1 | 3 | 229 | ||
NM_030927.2 | NP_112189.2 | 6 | 598 | |||
XM_006717992.3 | XP_006718055.1 | 6 | 754 | |||
XM_011540219.2 | XP_011538521.1 | 8 | 788 | |||
XM_011540220.2 | XP_011538522.1 | 8 | 710 | |||
XM_011540221.2 | XP_011538523.1 | 7 | 572 | |||
XM_011540222.2 | XP_011538524.1 | 7 | 750 | |||
XM_017016707.1 | XP_016872196.1 | 8 | 746 | |||
XM_017016708.1 | XP_016872197.1 | 7 | 707 | |||
XM_017016709.1 | XP_016872198.1 | 7 | 755 | |||
XM_017016710.1 | XP_016872199.1 | 7 | 706 | |||
XM_017016711.1 | XP_016872200.1 | 7 | 571 | |||
XM_017016712.1 | XP_016872201.1 | 7 | 774 | |||
XM_017016713.1 | XP_016872202.1 | 6 | 594 | |||
GenBank mRNA | AF311903.1 | - | 6 | 563 | ||
AK295321.1 | - | 3 | 200 | |||
AL136638.1 | - | 6 | 569 | |||
BC002920.2 | - | 5 | 493 | 3954042 | ||
BC093016.1 | - | 6 | 598 | 30528940 | ||
BI918996.1 | - | 3 | 252 | |||
DC300676.1 | - | 3 | 201 |