Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128309.1 | NP_001121781.1 | 5, 6 | 471 | ||
NM_030927.2 | NP_112189.2 | 8, 9 | 840 | |||
XM_006717992.3 | XP_006718055.1 | 8, 9 | 996 | |||
XM_011540219.2 | XP_011538521.1 | 10, 11 | 1030 | |||
XM_011540220.2 | XP_011538522.1 | 10, 11 | 952 | |||
XM_011540221.2 | XP_011538523.1 | 9, 10 | 814 | |||
XM_011540222.2 | XP_011538524.1 | 9, 10 | 992 | |||
XM_017016707.1 | XP_016872196.1 | 10, 11 | 988 | |||
XM_017016708.1 | XP_016872197.1 | 9, 10 | 949 | |||
XM_017016709.1 | XP_016872198.1 | 9, 10 | 997 | |||
XM_017016710.1 | XP_016872199.1 | 9, 10 | 948 | |||
XM_017016711.1 | XP_016872200.1 | 9, 10 | 813 | |||
XM_017016712.1 | XP_016872201.1 | 9, 10 | 1016 | |||
XM_017016713.1 | XP_016872202.1 | 8, 9 | 836 | |||
GenBank mRNA | AF311903.1 | - | 8, 9 | 805 | ||
AK055330.1 | - | 1, 2 | 849 | |||
AK295321.1 | - | 5, 6 | 442 | |||
AL136638.1 | - | 8, 9 | 811 | |||
BC002920.2 | - | 7, 8 | 735 | 3954042 | ||
BC093016.1 | - | 8, 9 | 840 | 30528940 | ||
BI918996.1 | - | 5, 6 | 494 | |||
DC300676.1 | - | 5, 6 | 443 |