Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128309.1 | NP_001121781.1 | 6 | 536 | ||
NM_030927.2 | NP_112189.2 | 9 | 905 | |||
XM_006717992.3 | XP_006718055.1 | 9 | 1061 | |||
XM_011540219.2 | XP_011538521.1 | 11 | 1095 | |||
XM_011540220.2 | XP_011538522.1 | 11 | 1017 | |||
XM_011540221.2 | XP_011538523.1 | 10 | 879 | |||
XM_011540222.2 | XP_011538524.1 | 10 | 1057 | |||
XM_017016707.1 | XP_016872196.1 | 11 | 1053 | |||
XM_017016708.1 | XP_016872197.1 | 10 | 1014 | |||
XM_017016709.1 | XP_016872198.1 | 10 | 1062 | |||
XM_017016710.1 | XP_016872199.1 | 10 | 1013 | |||
XM_017016711.1 | XP_016872200.1 | 10 | 878 | |||
XM_017016712.1 | XP_016872201.1 | 10 | 1081 | |||
XM_017016713.1 | XP_016872202.1 | 9 | 901 | |||
GenBank mRNA | AF311903.1 | - | 9 | 870 | ||
AK055330.1 | - | 2 | 914 | |||
AK295321.1 | - | 6 | 507 | |||
BC002920.2 | - | 8 | 800 | 3954042 | ||
BC093016.1 | - | 9 | 905 | 30528940 | ||
BI918996.1 | - | 6 | 559 | |||
DC300676.1 | - | 6 | 508 |