Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303043.1 | NP_001289972.1 | 4, 5 | 1639 | ||
NM_030935.4 | NP_112197.1 | 4, 5 | 1742 | |||
NR_130118.1 | 4, 5 | 897 | ||||
NR_130119.1 | 4, 5 | 894 | ||||
GenBank mRNA | AJ133115.1 | - | 4, 5 | 1327 | ||
AK296580.1 | - | 4, 5 | 1190 | |||
AK297702.1 | - | 4, 5 | 832 | |||
AK314824.1 | - | 4, 5 | 1537 | |||
BC001486.2 | - | 4, 5 | 1742 | 2966929 | ||
BC001966.1 | - | 4, 5 | 1595 | 3457297 | ||
BC002972.1 | - | 4, 5 | 1649 | 3538175 | ||
BC010406.2 | - | 4, 5 | 1656 | 3832780 |