Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052841.3 | NP_443073.1 | 2 | 797 | ||
GenBank mRNA | AF296450.1 | - | 2 | 459 | ||
AY048701.1 | - | 1 | 1199 | |||
BC035354.1 | - | 2 | 861 | 5171736 |
Gene Symbol: FAM229A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125095.1 | - | 1 | 415 |