Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160223.1 | NP_001153695.1 | 7 | 1081 | ||
NM_001160225.1 | NP_001153697.1 | 7 | 730 | |||
NM_030954.3 | NP_112216.3 | 7 | 851 | |||
NR_027668.1 | 6 | 1007 | ||||
NR_027669.1 | 6 | 777 | ||||
XM_006716404.2 | XP_006716467.1 | 8 | 838 | |||
XM_006716405.2 | XP_006716468.1 | 5 | 597 | |||
XM_011544666.2 | XP_011542968.1 | 7 | 635 | |||
XM_017013881.1 | XP_016869370.1 | 8 | 778 | |||
XM_017013882.1 | XP_016869371.1 | 6 | 623 | |||
GenBank mRNA | AF209504.1 | - | 2 | 188 | ||
AK027748.1 | - | 7 | 767 | |||
AK090864.1 | - | 6 | 940 | |||
AK095625.1 | - | 1 | 279 | |||
AK225086.1 | - | 7 | 731 | |||
AK225568.1 | - | 7 | 747 | |||
AL136620.1 | - | 7 | 735 | |||
BC013422.1 | - | 6 | 642 | |||
BC039461.1 | - | 7 | 838 | |||
BC058289.1 | - | 7 | 714 |