Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146008.1 | NP_001139480.1 | 4 | 2123 | ||
NM_030958.2 | NP_112220.2 | 5 | 2113 | |||
XM_017013883.1 | XP_016869372.1 | 5 | 2113 | |||
XM_017013884.1 | XP_016869373.1 | 5 | 2113 | |||
XM_017013885.1 | XP_016869374.1 | 4 | 1756 | |||
GenBank mRNA | AF205075.1 | - | 5 | 2022 | ||
AK022760.1 | - | 4 | 2123 | |||
BC137424.1 | - | 4 | 1908 | 9021422 |