Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170649.1 | NP_001164120.1 | 11 | 1635 | ||
NM_001170650.1 | NP_001164121.1 | 10 | 1509 | |||
NM_031206.4 | NP_112483.1 | 12 | 1686 | |||
XM_005262301.1 | XP_005262358.1 | 12 | 1671 | |||
XM_011531045.1 | XP_011529347.1 | 11 | 1535 | |||
XM_017029877.1 | XP_016885366.1 | 11 | 1620 | |||
XM_017029879.1 | XP_016885368.1 | 11 | 1561 | |||
XM_017029880.1 | XP_016885369.1 | 10 | 1510 | |||
GenBank mRNA | AF116730.1 | - | 12 | 1624 | ||
AK022587.1 | - | 12 | 1636 | |||
AK054817.1 | - | 2 | 1302 | |||
AK074087.1 | - | 11 | 4403 | |||
AK097383.1 | - | 10 | 1459 | |||
AK125760.1 | - | 1 | 1163 | |||
AK127207.1 | - | 1 | 691 | |||
AK225059.1 | - | 11 | 1568 | |||
BC014545.1 | - | 12 | 1670 | 4079376 | ||
BC018610.1 | - | 11 | 1583 | 3140243 | ||
BC019302.1 | - | 11 | 1580 | 2822893 |