Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013437.1 | NP_001013455.1 | 8 | 1132 | ||
NM_031216.3 | NP_112493.2 | 8 | 1132 | |||
XM_005258152.4 | XP_005258209.1 | 7 | 975 | |||
XM_011525742.2 | XP_011524044.1 | 9 | 1188 | |||
XM_011525743.2 | XP_011524045.1 | 8 | 1035 | |||
XM_011525744.1 | XP_011524046.1 | 9 | 1188 | |||
XM_011525745.2 | XP_011524047.1 | 8 | 956 | |||
XM_011525746.2 | XP_011524048.1 | 8 | 1238 | |||
XM_017026025.1 | XP_016881514.1 | 7 | 981 | |||
XM_017026026.1 | XP_016881515.1 | 7 | 1052 | |||
XM_017026027.1 | XP_016881516.1 | 8 | 1302 | |||
GenBank mRNA | AF136976.1 | - | 8 | 1100 | ||
AF255625.1 | - | 8 | 1036 | |||
AF431970.1 | - | 8 | 1036 | |||
AF514996.1 | - | 8 | 996 | |||
AK056940.1 | - | 8 | 1095 | |||
AK226075.1 | - | 5 | 3522 | |||
AK291226.1 | - | 8 | 1095 | |||
AK314403.1 | - | 8 | 1106 | |||
BC012430.1 | - | 8 | 1086 | 3865994 | ||
BX538037.1 | - | 3 | 2841 |