Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005088.2 | NP_005079.2 | 2 | 337 | ||
NR_027383.1 | 2 | 337 | ||||
GenBank mRNA | AK308367.1 | - | 2 | 332 | ||
BC016935.1 | - | 2 | 337 | |||
BC028151.1 | - | 2 | 305 | 5217247 | ||
BC110496.1 | - | 2 | 335 | 40007662 | ||
BC110497.1 | - | 2 | 335 | 40007663 | ||
L03426.1 | - | 2 | 307 | |||
M99578.1 | - | 2 | 310 |