Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005089.3 | NP_005080.1 | 1 | 67 | ||
XM_005274597.2 | XP_005274654.2 | 1 | 46 | |||
XM_011545590.2 | XP_011543892.2 | 1 | 795 | |||
XM_017029881.1 | XP_016885370.1 | 1 | 34 | |||
GenBank mRNA | AK309673.1 | - | 1 | 34 | ||
BC050451.1 | - | 1 | 38 | 6499970 | ||
BC065719.1 | - | 1 | 38 | |||
BC113454.1 | - | 1 | 67 | 8322506 | ||
BC113480.1 | - | 1 | 67 | 8322532 | ||
D49677.1 | - | 1 | 46 |