Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004651.3 | NP_004642.2 | 4, 5 | 659 | ||
XM_005272674.3 | XP_005272731.1 | 4, 5 | 496 | |||
XM_011543988.1 | XP_011542290.1 | 4, 5 | 453 | |||
GenBank mRNA | AB073597.1 | - | 4, 5 | 561 | ||
AK055588.1 | - | 3, 4 | 716 | |||
AK098394.1 | - | 4, 5 | 563 | |||
AK294635.1 | - | 4, 5 | 405 | |||
BC000350.2 | - | 4, 5 | 533 | |||
BC063668.1 | - | 4, 5 | 541 | |||
BC140849.1 | - | 4, 5 | 708 | 9021717 | ||
U44839.1 | - | 4, 5 | 519 |