Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256119.1 | NP_001243048.1 | 7 | 740 | ||
NM_001256120.1 | NP_001243049.1 | 8 | 767 | |||
NM_003491.3 | NP_003482.1 | 8 | 785 | |||
GenBank mRNA | AY183134.1 | - | 7 | 846 | ||
AY183135.1 | - | 7 | 601 | |||
BC000308.2 | - | 8 | 646 | 2820955 | ||
BC019312.1 | - | 8 | 646 | 2820955 | ||
BC063377.1 | - | 7 | 594 | 3050078 | ||
BI192945.1 | - | 8 | 582 | |||
CA313208.1 | - | 1 | 348 | |||
X77588.1 | - | 8 | 630 |