Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005677.3 | NP_005668.2 | 2 | 260 | ||
NM_080538.2 | NP_536799.1 | 2 | 389 | |||
NM_080539.3 | NP_536800.2 | 2 | 260 | |||
GenBank mRNA | AF057036.1 | - | 2 | 260 | ||
AJ225895.1 | - | 2 | 253 | |||
AK128401.1 | - | 2 | 389 | |||
AK289688.1 | - | 2 | 279 | |||
AY150334.1 | - | 2 | 237 | |||
AY150335.1 | - | 2 | 237 | |||
AY150336.1 | - | 2 | 237 | |||
AY150337.1 | - | 2 | 237 | |||
AY150338.1 | - | 2 | 237 | |||
AY150339.1 | - | 2 | 237 | |||
BC074828.2 | - | 2 | 164 | 30915542 | ||
BC074829.2 | - | 2 | 164 | 30915325 | ||
BX500613.1 | - | 2 | 247 |