Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031290.2 | NP_112580.2 | 2 | 956 | ||
GenBank mRNA | AK098719.1 | - | 2 | 956 | ||
AK225745.1 | - | 2 | 929 | |||
AL136887.1 | - | 2 | 927 | |||
BC069691.1 | - | 2 | 799 | 7262643 | ||
BC069748.1 | - | 2 | 799 | 7262655 | ||
BC069770.1 | - | 2 | 799 | 7262572 | ||
BC069806.1 | - | 2 | 799 | 7262584 |