Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014764.2 | NP_001014764.1 | 2 | 558 | ||
NM_031298.3 | NP_112588.1 | 2 | 445 | |||
GenBank mRNA | AA161105.1 | - | 1 | 303 | ||
BC001409.1 | - | 2 | 353 | 3139577 | ||
BM825446.1 | - | 2 | 339 | |||
BQ277705.1 | - | 2 | 459 | |||
BU931404.1 | - | 2 | 459 | |||
HY014822.1 | - | 2 | 446 |
Gene Symbol: P2RX5-TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037928.1 | 12 | 4783 | |||
GenBank mRNA | AB209623.1 | - | 12 | 4571 |