Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM167A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053279.2 | NP_444509.2 | 2 | 638 | ||
XM_005272398.4 | XP_005272455.1 | 3 | 991 | |||
XM_011543837.1 | XP_011542139.1 | 2 | 719 | |||
XM_011543838.2 | XP_011542140.1 | 2 | 645 | |||
XM_011543840.2 | XP_011542142.1 | 2 | 535 | |||
GenBank mRNA | AJ301564.1 | - | 2 | 547 | ||
AK290704.1 | - | 2 | 719 | |||
AL522457.3 | - | 2 | 638 | |||
AL834122.1 | - | 2 | 627 | |||
BC104041.1 | - | 1 | 460 | 40026945 | ||
BC104042.1 | - | 1 | 460 | 40026950 | ||
BC104043.1 | - | 1 | 460 | 40026951 |