Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031474.2 | NP_113662.1 | 6 | 885 | ||
XM_005253797.3 | XP_005253854.1 | 6 | 1036 | |||
XM_011521031.2 | XP_011519333.1 | 7 | 827 | |||
XM_017020000.1 | XP_016875489.1 | 4 | 1458 | |||
GenBank mRNA | AK054740.1 | - | 2 | 96 | ||
AK098063.1 | - | 6 | 856 | |||
AK300944.1 | - | 7 | 821 | |||
AL136557.1 | - | 6 | 853 | |||
BC036063.1 | - | 6 | 854 | 5276999 | ||
BC131562.1 | - | 6 | 842 | 40109200 |