Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300948.1 | NP_001287877.1 | 13 | 2324 | ||
NM_031490.3 | NP_113678.2 | 14 | 2456 | |||
GenBank mRNA | AJ548761.1 | - | 14 | 2323 | ||
AK027544.1 | - | 14 | 2289 | |||
AK027666.1 | - | 10 | 1491 | |||
AK074775.1 | - | 14 | 2321 | |||
AK090452.1 | - | 1 | 1257 | |||
AK127125.1 | - | 14 | 2088 | |||
AK130091.1 | - | 3 | 333 | |||
AK172790.1 | - | 5 | 611 | |||
AL834201.1 | - | 13 | 1822 | |||
BC002916.2 | - | 2 | 292 | |||
BC004234.2 | - | 2 | 292 | |||
BC093910.1 | - | 14 | 2290 | 7939755 | ||
BC093912.1 | - | 14 | 2290 | 7939757 | ||
BC110434.1 | - | 10 | 1472 | |||
BC143246.1 | - | 13 | 2158 | 9051747 |