Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031955.5 | NP_114161.3 | 5 | 1047 | ||
XM_006713778.2 | XP_006713841.1 | 5 | 1021 | |||
XM_017007308.1 | XP_016862797.1 | 5 | 1021 | |||
GenBank mRNA | AK093041.1 | - | 5 | 995 | ||
BC034496.2 | - | 5 | 949 | 4825905 |