Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122979.2 | NP_001116451.1 | 6 | 900 | ||
NM_001170631.1 | NP_001164102.1 | 5 | 983 | |||
NM_032029.4 | NP_114418.2 | 5 | 983 | |||
XM_011510051.2 | XP_011508353.1 | 6 | 932 | |||
XM_011510052.2 | XP_011508354.1 | 5 | 693 | |||
XM_017002442.1 | XP_016857931.1 | 6 | 928 | |||
GenBank mRNA | AF354295.1 | - | 3 | 432 | ||
AK298394.1 | - | 5 | 959 | |||
AY063125.1 | - | 3 | 366 | |||
BC109263.1 | - | 5 | 507 | |||
BC109264.2 | - | 5 | 507 |