Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142448.1 | NP_001135920.1 | 9 | 1281 | ||
NM_001142449.1 | NP_001135921.1 | 6 | 1296 | |||
NM_001142450.1 | NP_001135922.1 | 9 | 1431 | |||
NM_001142451.1 | NP_001135923.1 | 7 | 1362 | |||
NM_032038.2 | NP_114427.1 | 8 | 1518 | |||
XM_006721096.3 | XP_006721159.1 | 8 | 1132 | |||
XM_017023757.1 | XP_016879246.1 | 9 | 1304 | |||
XM_017023758.1 | XP_016879247.1 | 9 | 1300 | |||
XM_017023759.1 | XP_016879248.1 | 9 | 1284 | |||
GenBank mRNA | AF212371.1 | - | 8 | 1465 | ||
AF370423.1 | - | 9 | 1413 | |||
AL390215.1 | - | 6 | 1258 | |||
BC006156.1 | - | 8 | 1129 | |||
BC008325.1 | - | 8 | 1343 | 3501826 | ||
BC038961.1 | - | 9 | 1295 | 6053095 | ||
BC047741.1 | - | 7 | 1336 | 5757339 | ||
BC065235.1 | - | 9 | 1263 | 6161778 | ||
CD368853.1 | - | 4, 5 | 709 |