Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184957.1 | NP_001171886.1 | 17 | 2619 | ||
NM_001184958.1 | NP_001171887.1 | 17 | 2724 | |||
NM_032137.4 | NP_115513.4 | 17 | 3121 | |||
XM_006713341.2 | XP_006713404.1 | 18 | 3250 | |||
XM_006713342.2 | XP_006713405.1 | 18 | 3103 | |||
XM_011534153.2 | XP_011532455.1 | 18 | 4880 | |||
XM_011534154.2 | XP_011532456.1 | 18 | 4315 | |||
XM_011534157.2 | XP_011532459.1 | 14 | 2027 | |||
GenBank mRNA | AK131481.1 | - | 17 | 3916 | ||
AL136781.1 | - | 17 | 2539 | |||
AL834386.1 | - | 17 | 3073 | |||
BC030599.2 | - | 17 | 2682 | 4831210 | ||
BC038406.1 | - | 17 | 3041 | 5167835 |