Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326578.1 | NP_001313507.1 | 7 | 849 | ||
NM_001326579.1 | NP_001313508.1 | 7 | 636 | |||
NM_001326580.1 | NP_001313509.1 | 7 | 772 | |||
NM_003569.2 | NP_003560.2 | 7 | 772 | |||
NR_137169.1 | 7 | 772 | ||||
XM_011536178.1 | XP_011534480.1 | 7 | 688 | |||
GenBank mRNA | AK294911.1 | - | 7 | 521 | ||
BC011975.1 | - | 7 | 636 | 3860059 | ||
U77942.1 | - | 7 | 601 |